Learn More
Cayman Chemical N-Oleoyl Valine, Purity: ≥98%, CAS: 60374-41-6, 50 mg
SDP
Catalog No. 502984118
Encompass_Preferred
An endogenous N-acyl amine; antagonist at TRPV3; increases after acute lung injury in mouse lung tissue
Catalog No. 50-298-4118 Supplier Cayman Chemical Supplier No. 2006550 MG
An endogenous N-acyl amine; antagonist at TRPV3; increases after acute lung injury in mouse lung tissue