Learn More
Medchemexpress LLC N-Oleoyldopamine (OLDA) | 105955-11-1 | 99.5% | 417.62 | 1 ML
SDP
Catalog No. 5000347476
Encompass_Preferred
N-Oleoyldopamine (OLDA) is an endogenous TRPV1 selective agonist formed by the condensation of oleic acid and dopamine (DA). It can cross the blood-brain barrier and has been shown to protect the heart against ischemia-reperfusion injury through TRPV1 activation.
  • Endogenous TRPV1 selective agonist.
  • Crosses the blood-brain barrier.
  • Protects the heart against ischemia-reperfusion injury via activation of TRPV1.
Catalog No. 50-003-47476 Supplier Medchemexpress LLC Supplier No. HY10844810MM/1ML
N-Oleoyldopamine (OLDA) is an endogenous TRPV1 selective agonist formed by the condensation of oleic acid and dopamine (DA). It can cross the blood-brain barrier and has been shown to protect the heart against ischemia-reperfusion injury through TRPV1 activation.
  • Endogenous TRPV1 selective agonist.
  • Crosses the blood-brain barrier.
  • Protects the heart against ischemia-reperfusion injury via activation of TRPV1.