Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-oleoylethanolamide-d4 | 946524-36-3 | 99.0% | 329.55 | C20H35D4NO2 | 1 MG
SDP

Catalog No. 5000240653
Encompass_Preferred

Oleoylethanolamide-d4 is the deuterium-labeled analog of oleoylethanolamide used as a stable isotope reference standard and tracer in biochemical and metabolic research. Supplied in small, high-purity solution quantities, it supports analytical quantification, tracer studies, and method development.

  • High purity suitable for analytical use.
  • Deuterium-labeled internal standard for tracer and quantitation studies.
  • Molecular weight 329.55 and formula C20H35D4NO2.
  • Supplied as a solution for ease of handling and accurate dosing.
  • Recommended storage at -20°C for up to two years.

Catalog No. 50-002-40653 Supplier Medchemexpress LLC Supplier No. HY107542S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleoylethanolamide-d4 is the deuterium-labeled analog of oleoylethanolamide used as a stable isotope reference standard and tracer in biochemical and metabolic research. Supplied in small, high-purity solution quantities, it supports analytical quantification, tracer studies, and method development.

  • High purity suitable for analytical use.
  • Deuterium-labeled internal standard for tracer and quantitation studies.
  • Molecular weight 329.55 and formula C20H35D4NO2.
  • Supplied as a solution for ease of handling and accurate dosing.
  • Recommended storage at -20°C for up to two years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.