Learn More
Medchemexpress LLC N-omega-propyl-L-arginine | 137361-05-8 | 98.6% | 216.28 | C9H20N4O2 | 50 MG
SDP
Catalog No. 5000248463
Encompass_Preferred
N-omega-propyl-L-arginine is a research reagent and an amino acid analogue that acts as a potent, competitive, and selective inhibitor of neuronal nitric oxide synthase (nNOS). It is supplied as a solid with high purity for biochemical and pharmacological research applications.
  • Potent nNOS inhibitor with Ki = 57 nM.
  • High selectivity for nNOS (~149-fold over eNOS).
  • Research-grade purity (≈98.6%).
  • Solid form, appearance white to yellow.
  • Molecular weight 216.28 g/mol; CAS 137361-05-8.
Catalog No. 50-002-48463 Supplier Medchemexpress LLC Supplier No. HY10206250MG
N-omega-propyl-L-arginine is a research reagent and an amino acid analogue that acts as a potent, competitive, and selective inhibitor of neuronal nitric oxide synthase (nNOS). It is supplied as a solid with high purity for biochemical and pharmacological research applications.
  • Potent nNOS inhibitor with Ki = 57 nM.
  • High selectivity for nNOS (~149-fold over eNOS).
  • Research-grade purity (≈98.6%).
  • Solid form, appearance white to yellow.
  • Molecular weight 216.28 g/mol; CAS 137361-05-8.