Learn More
Enzo Life Sciences N-omega-Propyl-L-arginine (25mg). CAS: 137361-05-8
SDP
Catalog No. 502008424
Encompass_Preferred
Potent and selective inhibitor of neuronal nitric oxide synthase (nNOS; NOS I) relative to inducible nitric oxide synthase (iNOS; NOS II) (3158-fold) and endothelial nitric oxide synthase (eNOS; NOS III) (149-fold). Alternative name: N5-[Imino(propylamino)methyl]-L-ornithine. Formula: C9H20N4O2. MW: 216.3. Purity: ≥98% (TLC). Solubility: Soluble in PBS, pH 7.2 (25mg/ml), 100% ethanol (10mg/ml), DMSO (50μg/ml) or dimethyl formamide (50μg/ml). Use/Stability: Stable for at least one year when stored at -20°C. We do not recommend storing aqueous solutions for more than one day. Long Term Storage: -20°C.
Catalog No. 50-200-8424 Supplier Enzo Life Sciences Supplier No. ALX270203M025
Potent and selective inhibitor of neuronal nitric oxide synthase (nNOS; NOS I) relative to inducible nitric oxide synthase (iNOS; NOS II) (3158-fold) and endothelial nitric oxide synthase (eNOS; NOS III) (149-fold). Alternative name: N5-[Imino(propylamino)methyl]-L-ornithine. Formula: C9H20N4O2. MW: 216.3. Purity: ≥98% (TLC). Solubility: Soluble in PBS, pH 7.2 (25mg/ml), 100% ethanol (10mg/ml), DMSO (50μg/ml) or dimethyl formamide (50μg/ml). Use/Stability: Stable for at least one year when stored at -20°C. We do not recommend storing aqueous solutions for more than one day. Long Term Storage: -20°C.