Learn More
Medchemexpress LLC N-omega-propyl-L-arginine hydrochloride | 137361-05-8 | MFCD01320455 | >98.0% | 216.28 g/mol | C9H20N4O2 · xHCl | 25 MG
SDP
Catalog No. 5000248515
Encompass_Preferred
N-omega-propyl-L-arginine is a selective, competitive inhibitor of neuronal nitric oxide synthase (nNOS) used as a research reagent to study nitric oxide signaling and nNOS pharmacology. It is supplied as a white to off-white solid and is commonly provided in milligram quantities for laboratory use.
  • Selective nNOS inhibitor with reported Ki ≈ 57 nM.
  • High selectivity over eNOS (reported ~149-fold).
  • High purity suitable for research (reported ≥98% by HPLC).
  • Available in small-mass packages for research use (e.g., 25 mg).
  • Recommended storage: -20°C, desiccated to preserve stability.
Catalog No. 50-002-48515 Supplier Medchemexpress LLC Supplier No. HY10206225MG
N-omega-propyl-L-arginine is a selective, competitive inhibitor of neuronal nitric oxide synthase (nNOS) used as a research reagent to study nitric oxide signaling and nNOS pharmacology. It is supplied as a white to off-white solid and is commonly provided in milligram quantities for laboratory use.
  • Selective nNOS inhibitor with reported Ki ≈ 57 nM.
  • High selectivity over eNOS (reported ~149-fold).
  • High purity suitable for research (reported ≥98% by HPLC).
  • Available in small-mass packages for research use (e.g., 25 mg).
  • Recommended storage: -20°C, desiccated to preserve stability.