Learn More
Medchemexpress LLC N-omega-propyl-L-arginine hydrochloride | 2321366-46-3 | MFCD01320455 | 98.3% | 252.74 g/mol | C9H21ClN4O2 | 10 MG
SDP
Catalog No. 5000245922
Encompass_Preferred
N-omega-propyl-L-arginine hydrochloride is a potent, competitive, and highly selective inhibitor of neuronal nitric oxide synthase (nNOS), supplied as a hydrochloride salt for biochemical and pharmacological research. It is typically used in vitro to probe nitric oxide synthase function and selectivity.
  • Potent, competitive inhibitor of neuronal nitric oxide synthase (nNOS) with a reported Ki of 57 nM.
  • Shows 149-fold selectivity for nNOS over endothelial NOS (eNOS).
  • Supplied as the hydrochloride salt; formula C9H21ClN4O2 and molecular weight 252.74 g/mol.
  • Provided in milligram-scale quantities suitable for in vitro studies.
  • High purity confirmed by certificate of analysis (≈98.3% by LCMS).
Catalog No. 50-002-45922 Supplier Medchemexpress LLC Supplier No. HY102062A10MG
N-omega-propyl-L-arginine hydrochloride is a potent, competitive, and highly selective inhibitor of neuronal nitric oxide synthase (nNOS), supplied as a hydrochloride salt for biochemical and pharmacological research. It is typically used in vitro to probe nitric oxide synthase function and selectivity.
  • Potent, competitive inhibitor of neuronal nitric oxide synthase (nNOS) with a reported Ki of 57 nM.
  • Shows 149-fold selectivity for nNOS over endothelial NOS (eNOS).
  • Supplied as the hydrochloride salt; formula C9H21ClN4O2 and molecular weight 252.74 g/mol.
  • Provided in milligram-scale quantities suitable for in vitro studies.
  • High purity confirmed by certificate of analysis (≈98.3% by LCMS).