Learn More
Cayman Chemical N-Oxalylglycine, Purity: ≥98%, CAS: 5262-39-5, 100 mg
SDP
Catalog No. 502954082
Encompass_Preferred
A cell permeable inhibitor of α-ketoglutarate-dependent enzymes, including JMJD2A, JMJD2C, and JMJD2E (IC50s = 250, 500, and 24 μM, respectively); inhibits the prolyl hydroxylase domain-containing proteins PHD1 and PHD2 with IC50 values of 2.1 and 5.6 μM, respectively
Catalog No. 50-295-4082 Supplier Cayman Chemical Supplier No. 13944100 MG
A cell permeable inhibitor of α-ketoglutarate-dependent enzymes, including JMJD2A, JMJD2C, and JMJD2E (IC50s = 250, 500, and 24 μM, respectively); inhibits the prolyl hydroxylase domain-containing proteins PHD1 and PHD2 with IC50 values of 2.1 and 5.6 μM, respectively