Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-oxodecyl meglumine | 85261-20-7 | >98.0% | 349.46 g/mol | C17H35NO6 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW04021710G

Encompass_Preferred

MEGA-10 (N-oxodecyl meglumine) is a nonionic surfactant and biochemical reagent intended for research use in life-science applications, supplied as a high-purity solid.

  • Nonionic surfactant used in biochemical assays and sample preparation.
  • High purity, greater than 98.0%.
  • Molecular formula C17H35NO6.
  • Molecular weight 349.46 g/mol.
  • Available in small lab pack sizes suitable for research use.

Catalog No. 50-002-09112


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.