Learn More
Medchemexpress LLC HY-118628 25mg Medchemexpress, N-(p-amylcinnamoyl) Anthranilic Acid CAS:110683-10-8 Purity:>98%
SDP
Catalog No. 501932531
Encompass_Preferred
Medchemexpress, HY-118628 25mg N-(p-amylcinnamoyl) Anthranilic Acid CAS:110683-10-8 N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is a broad spectrum Phospholipase A 2 (PLA 2 ) inhibitor and TRP channel blocker [1] [2] . N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is also an effective reversible inhibitor of calcium-activated chloride channels , has potential to treat arrhythmia [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2531 Supplier Medchemexpress LLC Supplier No. HY11862825MG
Medchemexpress, HY-118628 25mg N-(p-amylcinnamoyl) Anthranilic Acid CAS:110683-10-8 N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is a broad spectrum Phospholipase A 2 (PLA 2 ) inhibitor and TRP channel blocker [1] [2] . N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is also an effective reversible inhibitor of calcium-activated chloride channels , has potential to treat arrhythmia [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.