Learn More
Medchemexpress LLC HY-113216 10mg Medchemexpress, Asymmetric dimethylarginine CAS:30315-93-6 Purity:>98%
SDP
Catalog No. 501969232
Encompass_Preferred
Asymmetric dimethylarginine is an endogenous inhibitor of nitric oxide synthase (NOS), and functions as a marker of endothelial dysfunction in a number of pathological states. CAS# 30315-93-6
Catalog No. 50-196-9232 Supplier Medchemexpress LLC Supplier No. HY11321610MG
Asymmetric dimethylarginine is an endogenous inhibitor of nitric oxide synthase (NOS), and functions as a marker of endothelial dysfunction in a number of pathological states. CAS# 30315-93-6