Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-P-Chlorobenzoyltyramine, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022929 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2929 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2929 Supplier MilliporeSigma Supelco Supplier No. BP756
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets

TRUSTED_SUSTAINABILITY

Specifications

CAS 41859-57-8
Molecular Formula C15H14ClNO2
Linear Formula C15H14ClNO2
Quantity 25 mg
MDL Number MFCD00218513
Synonym 4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
Molecular Weight (g/mol) 275.73
Formula Weight 275.73
Grade BP
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.