Learn More
Cayman Chemical N-(p-Coumaroyl) Serotonin, Purity: ≥95%, CAS: 68573-24-0, 10 mg
SDP
Catalog No. 502968271
Encompass_Preferred
An antioxidative phenolic naturally found in plants, including safflower seed and millet grain; ameliorates atherosclerosis in vivo, reducing lesion area in mice and rabbits
Catalog No. 50-296-8271 Supplier Cayman Chemical Supplier No. 1779710 MG
An antioxidative phenolic naturally found in plants, including safflower seed and millet grain; ameliorates atherosclerosis in vivo, reducing lesion area in mice and rabbits