Learn More
TARGETMOL CHEMICALS INC N-p-Coumaroyl serotonin 10MG
SDP
Catalog No. 502876538
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-(p-Coumaroyl) Serotonin, a polyphenol isolated from the seeds of safflower, exhibits antioxidant, cardioprotective effects after ischemia, and antitumor activity. It may improve vascular distensibility and reduce cardiovascular risk. N-(p-Coumaroyl) Serotonin ameliorates atherosclerosis and distensibility of the aortic wall in vivo, and is commonly used in atherosclerosis research. Purity 98.78%
Catalog No. 50-287-6538 Supplier TARGETMOL CHEMICALS INC Supplier No. TN196910MG
Add to Cart
Add to Cart
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-(p-Coumaroyl) Serotonin, a polyphenol isolated from the seeds of safflower, exhibits antioxidant, cardioprotective effects after ischemia, and antitumor activity. It may improve vascular distensibility and reduce cardiovascular risk. N-(p-Coumaroyl) Serotonin ameliorates atherosclerosis and distensibility of the aortic wall in vivo, and is commonly used in atherosclerosis research. Purity 98.78%