Learn More
Medchemexpress LLC N-(p-Coumaroyl) Serotonin | 68573-24-0 | 99.0% | 25 MG
SDP
Catalog No. 5000342227
Encompass_Preferred
N-(p-Coumaroyl) Serotonin is a selective inhibitor of PDGFRβ, ERK1/2, and caspase-8 with antioxidant, anti-atherosclerotic, and anti-inflammatory properties. It inhibits glioblastoma cells (U251MG, T98G) and normal fibroblasts. This compound inhibits PDGF signaling by reducing receptor phosphorylation, blocks Ca2+ influx, and induces apoptosis by activating caspase-8. It may improve atherosclerosis and aortic wall enlargement, and is also used in the study of glioblastoma.
  • Selective inhibitor of PDGFRβ, ERK1/2, and caspase-8
  • Possesses antioxidant, anti-atherosclerotic, and anti-inflammatory properties
  • Inhibits glioblastoma cell growth
  • Inhibits PDGF signaling by reducing receptor phosphorylation
  • Blocks Ca2+ influx and induces apoptosis
  • May improve atherosclerosis and aortic wall enlargement
Catalog No. 50-003-42227 Supplier Medchemexpress LLC Supplier No. 25MGHY129440
N-(p-Coumaroyl) Serotonin is a selective inhibitor of PDGFRβ, ERK1/2, and caspase-8 with antioxidant, anti-atherosclerotic, and anti-inflammatory properties. It inhibits glioblastoma cells (U251MG, T98G) and normal fibroblasts. This compound inhibits PDGF signaling by reducing receptor phosphorylation, blocks Ca2+ influx, and induces apoptosis by activating caspase-8. It may improve atherosclerosis and aortic wall enlargement, and is also used in the study of glioblastoma.
  • Selective inhibitor of PDGFRβ, ERK1/2, and caspase-8
  • Possesses antioxidant, anti-atherosclerotic, and anti-inflammatory properties
  • Inhibits glioblastoma cell growth
  • Inhibits PDGF signaling by reducing receptor phosphorylation
  • Blocks Ca2+ influx and induces apoptosis
  • May improve atherosclerosis and aortic wall enlargement