Learn More
Medchemexpress LLC N-(p-Coumaroyl) Serotonin | 68573-24-0 | 99.0% | 50 MG
SDP
Catalog No. 5000341360
Encompass_Preferred
N-(p-Coumaroyl) Serotonin is a selective inhibitor of PDGFRβ, ERK1/2, and caspase-8, exhibiting antioxidant, anti-atherosclerotic, and anti-inflammatory properties. It inhibits glioblastoma cells and demonstrates anti-inflammatory activity in RAW264.7 cells. The compound works by inhibiting PDGF signaling, blocking Ca2+ influx, and inducing apoptosis, making it suitable for glioblastoma and atherosclerosis research.
  • Selective inhibitor of PDGFRβ, ERK1/2, and caspase-8
  • Possesses antioxidant, anti-atherosclerotic, and anti-inflammatory properties
  • Inhibits glioblastoma cells
  • Reduces PDGF signaling and blocks Ca2+ influx
  • Induces apoptosis through caspase-8 activation
  • Improves atherosclerosis and aortic wall enlargement
Catalog No. 50-003-41360 Supplier Medchemexpress LLC Supplier No. HY12944050MG
N-(p-Coumaroyl) Serotonin is a selective inhibitor of PDGFRβ, ERK1/2, and caspase-8, exhibiting antioxidant, anti-atherosclerotic, and anti-inflammatory properties. It inhibits glioblastoma cells and demonstrates anti-inflammatory activity in RAW264.7 cells. The compound works by inhibiting PDGF signaling, blocking Ca2+ influx, and inducing apoptosis, making it suitable for glioblastoma and atherosclerosis research.
  • Selective inhibitor of PDGFRβ, ERK1/2, and caspase-8
  • Possesses antioxidant, anti-atherosclerotic, and anti-inflammatory properties
  • Inhibits glioblastoma cells
  • Reduces PDGF signaling and blocks Ca2+ influx
  • Induces apoptosis through caspase-8 activation
  • Improves atherosclerosis and aortic wall enlargement