Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific [N-(p-Toluenesulfonyl)imino]phenyliodinane, 55962-05-5, MFCD24850077, 1g

Catalog No. NC1300356
Encompass_Preferred

Molecular Formula C13H14INO2S, Purity 95%, Molecular Weight 375.23

Catalog No. NC1300356 Supplier Matrix Scientific Supplier No. 1159781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Molecular Formula C13H14INO2S, Purity 95%, Molecular Weight 375.23

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.