Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(p-Toluenesulfonyl)-L-alanine, 98%, Thermo Scientific™

Catalog No. AAH6691906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6691906 5 g
AAH6691914 25 g
AAH6691922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6691906 Supplier Thermo Scientific Chemicals Supplier No. H6691906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99076-56-9
MDL Number MFCD02259513
Synonym Tos-Ala-OH; N-Tosyl-L-alanine

Specifications

Melting Point 132°C to 137°C
Color White
Quantity 5 g
Formula Weight 243.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(p-Toluenesulfonyl)-L-alanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.