Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(p-Toluoyl)glycine 98.0+%, TCI America™
SDP

Catalog No. T03135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03135G 5 g
T031325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T03135G Supplier TCI America Supplier No. T03135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27115-50-0
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD00020449
InChI Key NRSCPTLHWVWLLH-UHFFFAOYSA-N
Synonym 4-methylhippuric acid, n-4-methylbenzoyl glycine, p-toluric acid, p-methylhippuric acid, n-p-toluoyl glycine, glycine, n-4-methylbenzoyl, 4-methyl hippuric acid, 2-4-methylbenzamido acetic acid, unii-9qox0dsk6f, hippuric acid, p-methyl
PubChem CID 97479
ChEBI CHEBI:68552
IUPAC Name 2-[(4-methylbenzoyl)amino]acetic acid
SMILES CC1=CC=C(C=C1)C(=O)NCC(=O)O

Specifications

Melting Point 161°C
Color White
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Toluoyl)glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.