Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(p-Tolyl)-1-naphthylamine 96.0+%, TCI America™
SDP

Catalog No. T2201500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2201500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2201500MG Supplier TCI America Supplier No. T2201500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 634-43-5
Molecular Formula C17H15N
Molecular Weight (g/mol) 233.314
InChI Key RWYRKFWBKGQTLU-UHFFFAOYSA-N
Synonym 1-(p-Toluidino)naphthalene
PubChem CID 235835
IUPAC Name N-(4-methylphenyl)naphthalen-1-amine
SMILES CC1=CC=C(C=C1)NC2=CC=CC3=CC=CC=C32

Specifications

Melting Point 78°C
Boiling Point 360°C
Quantity 500 mg
Formula Weight 233.31
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Tolyl)-1-naphthylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.