Learn More
TARGETMOL CHEMICALS INC N-p-Tosyl-GPR-pNA acetate 25MG
SDP
Catalog No. 502879078
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-(p-Tosyl)-GPR-pNA acetate is a colorimetric substrate for thrombin that can act as a measure of thrombin activity.
Catalog No. 50-287-9078 Supplier TARGETMOL CHEMICALS INC Supplier No. T3695125MG
Also available in 1 mg, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-(p-Tosyl)-GPR-pNA acetate is a colorimetric substrate for thrombin that can act as a measure of thrombin activity.