Learn More
Cayman Chemical N- p-Tosyl-GPR-pNAacetate 25mg
SDP
Catalog No. 502937306
Encompass_Preferred
A colorimetric thrombin substrate; cleavage of the GPR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of thrombin activity
Catalog No. 50-293-7306 Supplier Cayman Chemical Supplier No. 2756825 MG
A colorimetric thrombin substrate; cleavage of the GPR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of thrombin activity