Learn More
Cayman Chemical N-(p-Tosyl)-GPR-pNA (acetate), Purity: ≥98%, CAS: 86890-95-1, 50 mg
SDP
Catalog No. 502937315
Encompass_Preferred
A colorimetric thrombin substrate; cleavage of the GPR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of thrombin activity
Catalog No. 50-293-7315 Supplier Cayman Chemical Supplier No. 2756850 MG
A colorimetric thrombin substrate; cleavage of the GPR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of thrombin activity