Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-p-tosylglycine | 1080-44-0 | MFCD00045898 | 98.2% | 229.26 g/mol | C9H11NO4S | 25 G
SDP

Catalog No. 5000208854
Encompass_Preferred

N-p-tosylglycine is a tosyl-protected derivative of glycine supplied as a laboratory research reagent for use in organic synthesis and biochemical studies. It is provided as a white to off-white powder with high purity and is intended for research use only.

  • Tosyl-protected glycine derivative
  • Chemical formula C9H11NO4S
  • Molecular weight 229.26 g/mol
  • Cas number 1080-44-0
  • Purity 98.2%
  • Available in 25 g and 100 g sizes
  • Intended for research use only

Catalog No. 50-002-08854 Supplier Medchemexpress LLC Supplier No. 25GHYW053699
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-p-tosylglycine is a tosyl-protected derivative of glycine supplied as a laboratory research reagent for use in organic synthesis and biochemical studies. It is provided as a white to off-white powder with high purity and is intended for research use only.

  • Tosyl-protected glycine derivative
  • Chemical formula C9H11NO4S
  • Molecular weight 229.26 g/mol
  • Cas number 1080-44-0
  • Purity 98.2%
  • Available in 25 g and 100 g sizes
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.