Learn More
Medchemexpress LLC HY-N2230 5mg Medchemexpress, N-p-trans-Coumaroyltyramine CAS:36417-86-4 Purity:>98%
SDP
Catalog No. 501970377
Encompass_Preferred
Medchemexpress, HY-N2230 5mg N-p-trans-Coumaroyltyramine CAS:36417-86-4 N-p-trans-Coumaroyltyramine is a cinnamoylphenethyl amide isolated from polygonum hyrcanicum, acts as an acetylcholinesterase (AChE) inhibitor with an an IC50 of 122 μM. N-p-trans-Coumaroyltyramine exhibits anti-trypanosomal activity with an IC50 of 13.3 µM for T. brucei rhodesiense. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0377 Supplier Medchemexpress LLC Supplier No. HYN22305MG
Medchemexpress, HY-N2230 5mg N-p-trans-Coumaroyltyramine CAS:36417-86-4 N-p-trans-Coumaroyltyramine is a cinnamoylphenethyl amide isolated from polygonum hyrcanicum, acts as an acetylcholinesterase (AChE) inhibitor with an an IC50 of 122 μM. N-p-trans-Coumaroyltyramine exhibits anti-trypanosomal activity with an IC50 of 13.3 µM for T. brucei rhodesiense. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.