Learn More
Medchemexpress LLC N-p-trans-Coumaroyltyramine | 36417-86-4 | 1 MG
SDP
Catalog No. 5000420770
Encompass_Preferred
N-p-trans-Coumaroyltyramine is a natural phenolic amide compound. It acts as an inhibitor of Acetylcholinesterase (AChE) and α-glucosidase, and exhibits anti-trypanosomal activity. This compound can be utilized in research for diseases such as Alzheimer's disease.
  • Acts as an inhibitor of Acetylcholinesterase (IC50: 122 μM)
  • Acts as an inhibitor of α-glucosidase (IC50: 2.7 μM)
  • Exhibits anti-trypanosomal activity against *T. brucei rhodesiense* (IC50: 13.3 μM)
  • Utilized in research concerning diseases such as Alzheimer's disease
Catalog No. 50-004-20770 Supplier Medchemexpress LLC Supplier No. HYN22301MG
N-p-trans-Coumaroyltyramine is a natural phenolic amide compound. It acts as an inhibitor of Acetylcholinesterase (AChE) and α-glucosidase, and exhibits anti-trypanosomal activity. This compound can be utilized in research for diseases such as Alzheimer's disease.
  • Acts as an inhibitor of Acetylcholinesterase (IC50: 122 μM)
  • Acts as an inhibitor of α-glucosidase (IC50: 2.7 μM)
  • Exhibits anti-trypanosomal activity against *T. brucei rhodesiense* (IC50: 13.3 μM)
  • Utilized in research concerning diseases such as Alzheimer's disease