Learn More
TARGETMOL CHEMICALS INC N-p-trans-Coumaroyltyramine
SDP
Catalog No. 502877201
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-p-trans-Coumaroyltyramine is a natural product and an acetylcholinesterase (AChE) inhibitor with an IC50 of 122 uM. It also exhibits anti-trypanosomal activity with an IC50 of 13.3 uM for T. brucei rhodesiense. Purity 99.56%
Catalog No. 50-287-7201 Supplier TARGETMOL CHEMICALS INC Supplier No. TN10305MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-p-trans-Coumaroyltyramine is a natural product and an acetylcholinesterase (AChE) inhibitor with an IC50 of 122 uM. It also exhibits anti-trypanosomal activity with an IC50 of 13.3 uM for T. brucei rhodesiense. Purity 99.56%