Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-palmitoyl-D-erythro-sphingosine | 24696-26-2 | MFCD00056205 | >=99.0% | 537.90 g/mol | C34H67NO3 | 10 MG
SDP

Catalog No. 5000214351
Encompass_Preferred

C16-ceramide is a naturally occurring ceramide lipid used as a research reagent that activates p53 through direct and selective binding. It is provided in high purity for in vitro biochemical and cell-based studies.

  • Activates p53 through direct and selective binding.
  • High purity (≥99.0%) suitable for research use.
  • Molecular formula C34H67NO3; molecular weight 537.90 g/mol.
  • Available as a solid (multiple sizes including 10 mg) and as a 10 mM solution in ethanol.
  • Suitable for biochemical and cell-based studies of ceramide biology.
  • CAS number 24696-26-2 for unambiguous identification.

Catalog No. 50-002-14351 Supplier Medchemexpress LLC Supplier No. HY10035410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C16-ceramide is a naturally occurring ceramide lipid used as a research reagent that activates p53 through direct and selective binding. It is provided in high purity for in vitro biochemical and cell-based studies.

  • Activates p53 through direct and selective binding.
  • High purity (≥99.0%) suitable for research use.
  • Molecular formula C34H67NO3; molecular weight 537.90 g/mol.
  • Available as a solid (multiple sizes including 10 mg) and as a 10 mM solution in ethanol.
  • Suitable for biochemical and cell-based studies of ceramide biology.
  • CAS number 24696-26-2 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.