Learn More
Medchemexpress LLC N-Palmitoyl-D-sphingomyelin | 6254-89-3 | 99.9% | C39H79N2O6P | 1 MG
SDP
Catalog No. 5000353736
Encompass_Preferred
N-Palmitoyl-D-sphingomyelin is a component of LDL-carrying liposomes that can be used in colorectal cancer research. It has been shown to rescue cell death induction in HCT116 spheroids when combined with a respiratory chain inhibitor.
  • Component of LDL-carrying liposomes
  • Used in colorectal cancer research
  • Rescues cell death induction in HCT116 spheroids when combined with a respiratory chain inhibitor
Catalog No. 50-003-53736 Supplier Medchemexpress LLC Supplier No. HY1416291MG
N-Palmitoyl-D-sphingomyelin is a component of LDL-carrying liposomes that can be used in colorectal cancer research. It has been shown to rescue cell death induction in HCT116 spheroids when combined with a respiratory chain inhibitor.
  • Component of LDL-carrying liposomes
  • Used in colorectal cancer research
  • Rescues cell death induction in HCT116 spheroids when combined with a respiratory chain inhibitor