Learn More
Echelon Biosciences Research Labs N-PALMITOYL-DPPE 5mg
SDP
Catalog No. 5082241
Encompass
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-palmitoyl (N-palmitoyl-DPPE) is a type of N-acylphosphatidylethanolamine (NAPE) which as a group includes substrates in the endocannabinoid biosynthesis pathway. Fatty acid amides are formed by hydrolysis of NAPEs by a specific phospholiase D, and N-palmitoyl-ethanolamine (PEA) is found in significant concentrations in the mammalian circulation and central nervous systems. PEA is recognized for its anti-inflammatory and analgesic properties in cells and in vivo animal models of neuropathic and inflammatory diseases. Further, PEA-containing products are licensed for use in humans typically as an analgesic or anti-inflammatory with demonstrated high safety and tolerability. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-822-41 Supplier Echelon Biosciences Research Labs Supplier No. L24165MG
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-palmitoyl (N-palmitoyl-DPPE) is a type of N-acylphosphatidylethanolamine (NAPE) which as a group includes substrates in the endocannabinoid biosynthesis pathway. Fatty acid amides are formed by hydrolysis of NAPEs by a specific phospholiase D, and N-palmitoyl-ethanolamine (PEA) is found in significant concentrations in the mammalian circulation and central nervous systems. PEA is recognized for its anti-inflammatory and analgesic properties in cells and in vivo animal models of neuropathic and inflammatory diseases. Further, PEA-containing products are licensed for use in humans typically as an analgesic or anti-inflammatory with demonstrated high safety and tolerability. This item is non returnable