Learn More
TARGETMOL CHEMICALS INC N-Palmitoyl-L-aspartate 5MG
SDP
Catalog No. 502878211
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-Palmitoyl-L-aspartate (L-N-Palmitoylaspartic acid) is a surfactant used in detergents and is commonly used in the skin care industry. Purity 99.19%
Catalog No. 50-287-8211 Supplier TARGETMOL CHEMICALS INC Supplier No. T259185MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-Palmitoyl-L-aspartate (L-N-Palmitoylaspartic acid) is a surfactant used in detergents and is commonly used in the skin care industry. Purity 99.19%