Learn More
Medchemexpress LLC N-Palmitoyl-phenylalanine | 37571-96-3 | 99.7% | 403.60 | 5 G
SDP
Catalog No. 5000393514
Encompass_Preferred
N-Palmitoyl-phenylalanine is an N-acyl amide that contains the long-chain saturated fatty acid Palmitic acid conjugated via an amide bond to the essential amino acid L-Phenylalanine. It inhibits pre-mRNA splicing in HeLa cell nuclear lysates (IC50 > 400 μM) but has no splicing activity in intact HeLa cells.
  • An N-acyl amide derived from Palmitic acid and L-Phenylalanine.
  • Inhibits pre-mRNA splicing in HeLa cell nuclear lysates.
Catalog No. 50-003-93514 Supplier Medchemexpress LLC Supplier No. HY1697945G
Add to Cart
Add to Cart
N-Palmitoyl-phenylalanine is an N-acyl amide that contains the long-chain saturated fatty acid Palmitic acid conjugated via an amide bond to the essential amino acid L-Phenylalanine. It inhibits pre-mRNA splicing in HeLa cell nuclear lysates (IC50 > 400 μM) but has no splicing activity in intact HeLa cells.
  • An N-acyl amide derived from Palmitic acid and L-Phenylalanine.
  • Inhibits pre-mRNA splicing in HeLa cell nuclear lysates.