Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-palmitoyl taurine | 83982-06-3 | 99.0% | 10 MG
SDP

Catalog No. 5000234095
Encompass_Preferred

N-palmitoyl taurine is a fatty-acyl taurine endogenous metabolite supplied as a powder for biochemical and lipidomics research. The material is characterized by CAS 83982-06-3, molecular formula C18H37NO4S, and molecular weight 363.56.

  • Endogenous metabolite used in lipidomics and biochemical studies.
  • Supplied as a powder suitable for analytical and research applications.
  • Molecular formula C18H37NO4S; molecular weight 363.56.
  • Recommended storage: powder -20°C for up to 3 years, 4°C for up to 2 years.

Catalog No. 50-002-34095 Supplier Medchemexpress LLC Supplier No. HY12096510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-palmitoyl taurine is a fatty-acyl taurine endogenous metabolite supplied as a powder for biochemical and lipidomics research. The material is characterized by CAS 83982-06-3, molecular formula C18H37NO4S, and molecular weight 363.56.

  • Endogenous metabolite used in lipidomics and biochemical studies.
  • Supplied as a powder suitable for analytical and research applications.
  • Molecular formula C18H37NO4S; molecular weight 363.56.
  • Recommended storage: powder -20°C for up to 3 years, 4°C for up to 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.