Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-palmitoyl taurine | 83982-06-3 | 99.0% | 25 MG
SDP

Catalog No. 5000232794
Encompass_Preferred

N-palmitoyl taurine is an endogenous lipid metabolite supplied for research use. It is provided as a high-purity analytical standard suitable for biochemical and pharmacological studies.

  • High purity (99.0%).
  • CAS number 83982-06-3.
  • Molecular formula C18H37NO4S; molecular weight 363.56.
  • Soluble in ethanol (approx. 5 mg/mL with ultrasonic warming and pH adjustment).
  • Powder storage: -20°C for long-term (3 years) or 4°C (2 years); in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in small milligram-scale quantities including 25 mg.

Catalog No. 50-002-32794 Supplier Medchemexpress LLC Supplier No. HY12096525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-palmitoyl taurine is an endogenous lipid metabolite supplied for research use. It is provided as a high-purity analytical standard suitable for biochemical and pharmacological studies.

  • High purity (99.0%).
  • CAS number 83982-06-3.
  • Molecular formula C18H37NO4S; molecular weight 363.56.
  • Soluble in ethanol (approx. 5 mg/mL with ultrasonic warming and pH adjustment).
  • Powder storage: -20°C for long-term (3 years) or 4°C (2 years); in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in small milligram-scale quantities including 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.