Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-palmitoyl taurine | 83982-06-3 | 99.0% | 50 MG
SDP

Catalog No. 5000233989
Encompass_Preferred

N-palmitoyl taurine is an endogenous fatty-acyl taurine used as a research reagent in studies of lipid signaling, endocannabinoid biology, and metabolomics. It is supplied as a white to off-white solid and is commonly used as an analytical standard or biochemical probe in pharmacology and biochemical assays.

  • White to off-white solid form for laboratory use.
  • Purity 99.0% for analytical and research applications.
  • Molecular formula C18H37NO4S; molecular weight 363.6 g/mol.
  • Soluble in ethanol (≈5 mg/mL) with warming and acidification.
  • Store powder at -20°C for long-term stability; store solutions at -80°C.

Catalog No. 50-002-33989 Supplier Medchemexpress LLC Supplier No. HY12096550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-palmitoyl taurine is an endogenous fatty-acyl taurine used as a research reagent in studies of lipid signaling, endocannabinoid biology, and metabolomics. It is supplied as a white to off-white solid and is commonly used as an analytical standard or biochemical probe in pharmacology and biochemical assays.

  • White to off-white solid form for laboratory use.
  • Purity 99.0% for analytical and research applications.
  • Molecular formula C18H37NO4S; molecular weight 363.6 g/mol.
  • Soluble in ethanol (≈5 mg/mL) with warming and acidification.
  • Store powder at -20°C for long-term stability; store solutions at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.