Learn More
Cayman Chemical N-(p-amylcinnamoyl) Anthranilic Acid, Purity: ≥95%, CAS: 110683-10-8, 100 mg
SDP
Catalog No. 502968726
Encompass_Preferred
A channel blocker that acts on several TRP channels, including TRPM2, TRPM8, and TRPC6 (IC50 = 1.7, 3.8, and 2.3 μM, respectively); inhibits PLA2, blocking the release of arachidonic acid when given at 50 μM
Catalog No. 50-296-8726 Supplier Cayman Chemical Supplier No. 14531100 MG
A channel blocker that acts on several TRP channels, including TRPM2, TRPM8, and TRPC6 (IC50 = 1.7, 3.8, and 2.3 μM, respectively); inhibits PLA2, blocking the release of arachidonic acid when given at 50 μM