Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Paraffin Mix C7, C8, C9 & C10 Analytical Standard, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022538 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2538 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2538 Supplier MilliporeSigma Supelco Supplier No. 47101

missing translation for 'validMainframeMsg'

May be used for determining column resolution, retention times, and response factors.

TRUSTED_SUSTAINABILITY

Specifications

Flash Point -3.89°C (Closed Cup)
UN Number UN3295 - Class 3 - PG 2 - EHS - VP > 110 kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR)
Quantity 500 mg
Grade Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.