Learn More
Medchemexpress LLC N-(Peg2-Boc)-N-bis(peg2-propargyl) | 2100306-69-0 | 485.61 | 100 MG
SDP
Catalog No. 5000350310
Encompass_Preferred
N-(PEG2-Boc)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
Catalog No. 50-003-50310 Supplier Medchemexpress LLC Supplier No. HY140093100MG
N-(PEG2-Boc)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.