Learn More
Medchemexpress LLC N-(PEG2-Boc)-N-bis(PEG2-propargyl) | 2100306-69-0 | 485.61 | 50 MG
SDP
Catalog No. 5000351402
Encompass_Preferred
N-(PEG2-Boc)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.
  • It is a PEG-based PROTAC linker.
  • It can be used in the synthesis of PROTACs.
  • It is a click chemistry reagent.
  • It contains an alkyne group.
  • It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
Catalog No. 50-003-51402 Supplier Medchemexpress LLC Supplier No. HY14009350MG
N-(PEG2-Boc)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.
  • It is a PEG-based PROTAC linker.
  • It can be used in the synthesis of PROTACs.
  • It is a click chemistry reagent.
  • It contains an alkyne group.
  • It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.