Learn More
Medchemexpress LLC N-Pentadecanoyl-psychosine | 112726-51-9 | 98.0% | C39H75NO8 | 1 MG
SDP
Catalog No. 5000354739
Encompass_Preferred
N-Pentadecanoyl-psychosine is a glycolipid that can be used in membrane adhesion studies.
  • Glycolipid used in membrane adhesion studies
  • Provided as a white to off-white solid
  • Purity of 98.0%
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-54739 Supplier Medchemexpress LLC Supplier No. HY1429751MG
N-Pentadecanoyl-psychosine is a glycolipid that can be used in membrane adhesion studies.
  • Glycolipid used in membrane adhesion studies
  • Provided as a white to off-white solid
  • Purity of 98.0%
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month