Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003540 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-540 500 mL Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-540 Supplier Avantor Supplier No. T00707
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 109-66-0
Quantity 500 mL
Flash Point −40.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.63kg
Molecular Formula C5H12
MDL Number MFCD00009498
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES CCCCC
IUPAC Name pentane
Molecular Weight (g/mol) 72.15
PubChem CID 8003
ChEBI CHEBI:37830
Formula Weight 72.15
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material n-Pentane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.