Learn More
Medchemexpress LLC N-Phenethylbenzamide | 3278-14-6 | 99.6% | 225.29 | 5 G
SDP
Catalog No. 5000401298
Encompass_Preferred
N-Phenethylbenzamide is an active compound that can be extracted from *Liriodendron tulipifera*. It is suitable for research related to inflammatory diseases.
  • Inhibits LPS-induced NO production in RAW264.7 cells with an IC50 value of >50 μM.
Catalog No. 50-004-01298 Supplier Medchemexpress LLC Supplier No. HY321355G
N-Phenethylbenzamide is an active compound that can be extracted from *Liriodendron tulipifera*. It is suitable for research related to inflammatory diseases.
  • Inhibits LPS-induced NO production in RAW264.7 cells with an IC50 value of >50 μM.