Learn More
Strem, An Ascensus Company CAS# 740815-36-5. 2g. N-Phenyl-2-(dicyclohexylphosphino)indol, min. 95% [cataCXium PInCy]. MFCD06798303
SDP
Supplier:  Strem, An Ascensus Company 1536052G
Encompass
CAS# 740815-36-5. 2g. N-Phenyl-2-(dicyclohexylphosphino)indol, min. 95% [cataCXium PInCy]. MFCD06798303. Molecular Weight: 389.51. Molecular Formula: C26H32NP. Color/form: white to yellow pwdr. Strem# 15-3605. http://www.strem.com/catalog/v/15-3605/
Catalog No. 50-138-5418