Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N'-Phenylacetohydrazide | 114-83-0 | MFCD00008672 | 100.0% | 150.18 g/mol | C8H10N2O | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW017425

Encompass_Preferred

N'-Phenylacetohydrazide (CAS 114-83-0) is an organic reagent used in biochemical assays and organic synthesis, commonly for detecting and derivatizing carbonyl compounds such as aldehydes and ketones. It is supplied as a solid, off-white to light yellow powder with formula C8H10N2O and molecular weight 150.18 g/mol, offered at high purity for laboratory use.

  • High purity (99.99%).
  • Molecular weight 150.18 g/mol.
  • Chemical formula C8H10N2O.
  • Off-white to light yellow solid.
  • Common reagent for determination of aldehydes and ketones.
  • Soluble in DMSO and ethanol.

Catalog No. 50-003-03828


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.