Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Phenylacetyl-L-homoserine lactone | 55154-48-8 | >99.6% | 219.24 | 25 MG
SDP

Catalog No. 5000352201
Encompass_Preferred

N-Phenylacetyl-L-homoserine lactone is a small diffusible signaling molecule known for its strong antagonistic or super-activating effects on the quorum sensing of Vibrio fischeri. This high-purity chemical reagent is intended for research use only.

  • Purity of 99.60% confirmed by LCMS.
  • Molecular weight is 219.24.
  • Store as powder at -20°C for up to 3 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-003-52201 Supplier Medchemexpress LLC Supplier No. 25MGHY137407
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Phenylacetyl-L-homoserine lactone is a small diffusible signaling molecule known for its strong antagonistic or super-activating effects on the quorum sensing of Vibrio fischeri. This high-purity chemical reagent is intended for research use only.

  • Purity of 99.60% confirmed by LCMS.
  • Molecular weight is 219.24.
  • Store as powder at -20°C for up to 3 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.