Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-phenylacetyl-L-homoserine lactone | 55154-48-8 | C12H13NO3 | 10MG
SDP

Catalog No. 5000258999
Encompass_Preferred

N-Phenylacetyl-L-homoserine lactone is a small, diffusible bacterial signaling molecule used as a research reagent to modulate quorum sensing. It can antagonize or super-activate quorum sensing in Vibrio fischeri and is used to study bacterial communication and related anti-infection pathways.

  • Purity 99.6% (LCMS).
  • Molecular formula C12H13NO3 and molecular weight 219.24 g·mol⁻¹.
  • Soluble in DMSO at ≥125 mg/mL.
  • Store powder at -20°C (stable up to 3 years); in solution store at -80°C for up to 6 months or -20°C for 1 month.
  • Intended for use in quorum sensing and bacterial pathway research.

Catalog No. 50-002-58999 Supplier Medchemexpress LLC Supplier No. HY13740710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Phenylacetyl-L-homoserine lactone is a small, diffusible bacterial signaling molecule used as a research reagent to modulate quorum sensing. It can antagonize or super-activate quorum sensing in Vibrio fischeri and is used to study bacterial communication and related anti-infection pathways.

  • Purity 99.6% (LCMS).
  • Molecular formula C12H13NO3 and molecular weight 219.24 g·mol⁻¹.
  • Soluble in DMSO at ≥125 mg/mL.
  • Store powder at -20°C (stable up to 3 years); in solution store at -80°C for up to 6 months or -20°C for 1 month.
  • Intended for use in quorum sensing and bacterial pathway research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.