Learn More
Medchemexpress LLC N-phenylacetyl-L-homoserine lactone | 55154-48-8 | MFCD00075591 | >=98% | 219.24 | 1 MG
SDP
Catalog No. 5000349652
Encompass_Preferred
N-Phenylacetyl-L-homoserine lactone (PAHL) is a homoserine lactone derivative, which is a quorum sensing molecule produced by Gram-negative bacteria. It can be used for the research of antibacterial agents.
  • Quorum sensing (QS) is a process of cell-to-cell communication that allows bacteria to sense their population density and coordinate group behaviors.
  • Homoserine lactones (HSLs) are a class of QS signaling molecules used by Gram-negative bacteria.
  • It is a homoserine lactone derivative.
Catalog No. 50-003-49652 Supplier Medchemexpress LLC Supplier No. HY1374071MG
N-Phenylacetyl-L-homoserine lactone (PAHL) is a homoserine lactone derivative, which is a quorum sensing molecule produced by Gram-negative bacteria. It can be used for the research of antibacterial agents.
  • Quorum sensing (QS) is a process of cell-to-cell communication that allows bacteria to sense their population density and coordinate group behaviors.
  • Homoserine lactones (HSLs) are a class of QS signaling molecules used by Gram-negative bacteria.
  • It is a homoserine lactone derivative.