Learn More
Medchemexpress LLC N-phenylacrylamide | 2210-24-4 | MFCD00080617 | 99.9% | 147.17 g/mol | C9H9NO | 25 G
SDP
Catalog No. 5000303105
Encompass_Preferred
N-phenylacrylamide (CAS 2210-24-4) is a small-molecule research reagent used as an ochratoxin A binder for mycotoxin extraction and analytical workflows. It is supplied as a solid with high reported purity for laboratory assays.
  • High purity (reported ~99.9%), suitable for analytical use.
  • Acts as an ochratoxin A binder for mycotoxin extraction and cleanup.
  • Supplied as a solid, facilitating storage and handling in laboratories.
  • Molecular formula C9H9NO and molecular weight 147.17 g/mol.
  • Available in multiple pack sizes including 25 g for bulk use.
  • Manufacturer datasheet and safety data sheet available for handling and quality verification.
Catalog No. 50-003-03105 Supplier Medchemexpress LLC Supplier No. 25GHYW018582
N-phenylacrylamide (CAS 2210-24-4) is a small-molecule research reagent used as an ochratoxin A binder for mycotoxin extraction and analytical workflows. It is supplied as a solid with high reported purity for laboratory assays.
  • High purity (reported ~99.9%), suitable for analytical use.
  • Acts as an ochratoxin A binder for mycotoxin extraction and cleanup.
  • Supplied as a solid, facilitating storage and handling in laboratories.
  • Molecular formula C9H9NO and molecular weight 147.17 g/mol.
  • Available in multiple pack sizes including 25 g for bulk use.
  • Manufacturer datasheet and safety data sheet available for handling and quality verification.