Learn More
Medchemexpress LLC N-Phenylnaphthalen-1-amine | 90-30-2 | 100.0% | 219.29 | 100 G
SDP
Catalog No. 5000453386
Encompass_Preferred
N-Phenyl-1-naphthylamine is a dye that fluoresces strongly upon binding to the inner phospholipid bilayer of Gram-negative bacteria. This property makes it useful for measuring outer membrane permeability. It also serves as a fluorescence probe for odorant-binding proteins (OBP), demonstrating a dissociation constant of 1.67 μM. The compound exhibits an excitation wavelength of 337 nM and an emission wavelength of 407 nM.
  • Fluoresces strongly when bound to the inner phospholipid bilayer of Gram-negative bacteria.
  • Can be used to measure outer membrane permeability.
  • Functions as a fluorescence probe for odorant-binding proteins.
  • Exhibits an excitation wavelength of 337 nM.
  • Exhibits an emission wavelength of 407 nM.
Catalog No. 50-004-53386 Supplier Medchemexpress LLC Supplier No. HYW009756100G
N-Phenyl-1-naphthylamine is a dye that fluoresces strongly upon binding to the inner phospholipid bilayer of Gram-negative bacteria. This property makes it useful for measuring outer membrane permeability. It also serves as a fluorescence probe for odorant-binding proteins (OBP), demonstrating a dissociation constant of 1.67 μM. The compound exhibits an excitation wavelength of 337 nM and an emission wavelength of 407 nM.
  • Fluoresces strongly when bound to the inner phospholipid bilayer of Gram-negative bacteria.
  • Can be used to measure outer membrane permeability.
  • Functions as a fluorescence probe for odorant-binding proteins.
  • Exhibits an excitation wavelength of 337 nM.
  • Exhibits an emission wavelength of 407 nM.