Learn More
Medchemexpress LLC N-Phenylnaphthalen-1-amine | 90-30-2 | MFCD00003878 | 99.9% | 219.29 | 50 G
SDP
Catalog No. 5000453103
Encompass_Preferred
N-Phenylnaphthalen-1-amine is a dye that strongly fluoresces when bound to the inner phospholipid bilayer of Gram-negative bacteria and can be used to measure outer membrane permeability. It also functions as a fluorescence probe for odorant-binding proteins (OBP).
  • Used to measure outer membrane permeability of Gram-negative bacteria.
  • Functions as a fluorescence probe for odorant-binding proteins (OBP).
  • Exhibits an excitation wavelength of 337 nM and an emission wavelength of 407 nM.
Catalog No. 50-004-53103 Supplier Medchemexpress LLC Supplier No. HYW00975650G
N-Phenylnaphthalen-1-amine is a dye that strongly fluoresces when bound to the inner phospholipid bilayer of Gram-negative bacteria and can be used to measure outer membrane permeability. It also functions as a fluorescence probe for odorant-binding proteins (OBP).
  • Used to measure outer membrane permeability of Gram-negative bacteria.
  • Functions as a fluorescence probe for odorant-binding proteins (OBP).
  • Exhibits an excitation wavelength of 337 nM and an emission wavelength of 407 nM.